Merck
TMO140620
Nunc® Cell Culture Inserts in 24-well Nunclon Delta surface plate
cell culture treated, polycarbonate, sterile, pore size 0.4 μm, case of 48 ea (supplied as 4 x 24 well plates, each containing 12 inserts)
Synonyma:
multidish membrane insert, treated membrane insert, Nunc 140620, cultivation plate, surface treated dish
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
NACRES:
NB.13
UNSPSC Code:
41122102
Material:
polycarbonate
Sterility:
sterile
Binding type:
cell culture treated
Manufacturer/tradename:
Nunc 140620
Packaging:
case of 48 ea (supplied as 4 x 24 well plates, each containing 12 inserts)
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
polycarbonate
sterility
sterile
packaging
case of 48 ea (supplied as 4 x 24 well plates, each containing 12 inserts)
manufacturer/tradename
Nunc 140620
pore size
0.4 μm pore size
binding type
cell culture treated
General description
Cultivate most cell types without matrix coating when using Thermo Scientific Nunc Polycarbonate Membrane Inserts in Multidishes.
Application
  • Tumor migration and invasion assays.
  • Epithelial (e.g. Caco-2) and endothelial (e.g. brain microvascular endothelial cells) barriers assays.
  • Dermal/epidermal and epithelial tissue models (e.g. artificial skin model).
  • Chemotaxis studiesCo-culture.
Features and Benefits
  • Tissue culture-treated porous membrane for optimized cell attachment.
  • Three pore sizes: 0.4, 3, and 8 micron.
  • Pre-packed in sterile Nunclon™ Delta-treated multi-well plates.
  • Nontoxic and nonpyrogenic.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Still not finding the right product?
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)