Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
10 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 130,00 Kč |
O této položce
1 130,00 Kč
material
white polystyrene
sterility
non-sterile
feature
to hold size 00 to 2 tablets and capsules
packaging
pack of 10 ea
manufacturer/tradename
Heathrow Scientific 120846
L × W × H
10.3 in. × 5.2 in. × 0.5 in.
General description
For Capsule/Tablets size 00 to 2.
Available in both, sterile* and non-sterile.
Features and Benefits
- Disposable to ensure no chemical cross-contamination for patient safety
- Made from FDA food grade polystyrene in an ISO Class 8 clean room
- Meets quality control and GMP requirements for manufacturing batch sampling
- Durable and shatterproof trays constructed of inert, BSE/TSE free material
- Pour spout in corner for dispensing control
- Ergonomic spatula (length - 6.9 in/17.6 cm
1 of 1
Tato položka | |||
|---|---|---|---|
| packaging pack of 10 ea | packaging pack of 30 ea (3 x packs of 10 individually wrapped) | packaging pack of 10 ea | packaging pack of 30 ea (3 x packs of 10 ea) |
| material white polystyrene | material white polystyrene | material white polystyrene | material white polystyrene |
| manufacturer/tradename Heathrow Scientific 120846 | manufacturer/tradename Heathrow Scientific 120847 | manufacturer/tradename Heathrow Scientific 120845 | manufacturer/tradename Heathrow Scientific 120848 |
| sterility non-sterile | sterility sterile; electron beam irradiated | sterility sterile; electron beam irradiated | sterility non-sterile |
| feature to hold size 00 to 2 tablets and capsules | feature to hold round pills and capsules | feature to hold size 00 to 2 tablets and capsules | feature to hold round pills and capsules |
| L × W × H 10.3 in. × 5.2 in. × 0.5 in. | L × W × H 23.0 in. × 23.0 in. × 1.3 in. | L × W × H 10.3 in. × 5.2 in. × 0.5 in. | L × W × H 23.0 in. × 23.0 in. × 1.3 in. |
Still not finding the right product?
Explore all of our products under Rectangular Capsule/Tablet Counting Tray
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?