Merck
HS120250
Wide mouth safety labelled wash bottle
capacity 500 mL, label, Acetone, self venting, PE-LD
Synonyma:
Heathrow Scientific HS120250, cleaning bottle, dispensing bottle, drip bottle, drop bottle, fluid dispenser, laboratory wash bottle, solvent wash bottle, squeezable bottle, washbottle
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
NACRES:
NB.43
UNSPSC Code:
41103209
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
PE-LD , low-density polyethylene bottle, polypropylene cap, wide-mouth bottle, red cap
joint
joint threaded
feature
label Acetone, vented, with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120250
capacity
500 mL
General description
The information provided on the front of each bottle is specific to the product contained and includes the following:
  • Chemical Abstract Service (CAS) No.
  • National Fire Protection Association (NFPA) codes and symbols
  • Protective Equipment Symbol (PES)
  • Target Organ Information and Hazard Codes.
You also have the option of using other liquids within the bottles and creating the appropriate safety information on the outside of the bottle by using the ‘Self Labeling’ versions. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the floSelf Labeling Bottles must only be used with chemicals that are safe and compatible with LDPE
Application
Used for easy identification of solutions and solvents for use on the bench.
Features and Benefits
  • Wide bottle mouth makes filling easier
  • Non-metallic vent that prevents the risk of solvent leakage
  • Closure and stem are molded in one piece to make them leak-proof
  • Bottles are made from LDPE, with a PP closure with the exception of the bottle designed for Sodium Hypochlorite, which is a white LDPE bottle for light protection
  • Six named chemicals, a self labeling and assorted pack of bottles to choose from
  • 500 mL bottle size
  • Supplied in packs of 6
Still not finding the right product?
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)