Merck
GE18-1153-04
Tricorn Glass Tube
joint: 5/20
Synonyma:
18-1153-04, Cytiva accessory, FPLC spare part, chromatography tubing, empty column
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
NACRES:
NB.43
UNSPSC Code:
41115709
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
joint
joint: 5/20
manufacturer/tradename
Cytiva 18-1153-04
18-1153-13
General description
Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
Tricorn columns incorporate modern design and high performance to increase the user friendliness and reliability of the chromatography column. Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures.
  • High performance glass columns designed for reliable high resolution, reproducibility and durability
  • New longer adaptor for increased adjustment of bed height
  • Modern, integrated design – simple to assemble, pack, operate, and maintain
  • Broad range of column dimensions – suitable for many different purification needs
  • Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
  • Locking ring – prevents accidental compression of media bed
  • Separate packing connector – allows simple column packing for a broad range of media
Still not finding the right product?
Explore all of our products under Tricorn Glass Tube
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)