O této položce
NACRES:
NB.14
UNSPSC Code:
41122403
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlisterility
sterile
manufacturer/tradename
Corning 3007
L
250 mm
General description
Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® single use spatulas
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?