O této položce
NACRES:
NB.71
UNSPSC Code:
41100000
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
stainless steel
packaging
pack of 22 pieces
manufacturer/tradename
Benchmark Scientific W1105-22200
size
0.001-200 g
General description
Basic, compact balances with readability in the range of 0.1g to 0.01g can be calibrated with M1 Class weights.
Each individual weight and weight set from Accuris™ is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
Each individual weight and weight set from Accuris™ is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
The economy M1 Calibration Weight Set includes 22 pieces in a protective carrying case.
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
Still not finding the right product?
Explore all of our products under Accuris™ M1 Grade Calibration Weight Set
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?