Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
500 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 3 280,00 Kč |
O této položce
3 280,00 Kč
material
cap (screw, separate), clear tube, conical bottom, polypropylene tube
sterility
non-sterile
feature
DNase free, RNase free, graduations
packaging
pack of 500 ea (5 x bags 100ea)
manufacturer/tradename
Benchmark Scientific C1005-SC5
H
-64 mm
capacity
5 mL
tube diam.
16 mm
suitability
suitable for (centrifugation), suitable for PCR
General description
Application
1 of 1
Tato položka | |||
|---|---|---|---|
| sterility non-sterile | sterility sterile | sterility sterile | sterility sterile (non-pyrogenic) |
| material cap (screw, separate), conical bottom, clear tube, polypropylene tube | material black tube, cap (screw, attached), conical bottom, polypropylene tube | material cap (screw, attached), clear tube, conical bottom, polypropylene tube | material cap (plugseal), conical bottom tube, polypropylene |
| suitability suitable for (centrifugation), suitable for PCR | suitability suitable for (centrifugation), suitable for PCR | suitability suitable for (centrifugation), suitable for PCR | suitability suitable for (applications requiring large volume centrifugation) |
| manufacturer/tradename Benchmark Scientific C1005-SC5 | manufacturer/tradename Benchmark Scientific C1005-SC5-OB | manufacturer/tradename Benchmark Scientific C1005-SC5-S | manufacturer/tradename Corning 431123 |
| capacity 5 mL | capacity 5 mL | capacity 5 mL | capacity 500 mL |
| packaging pack of 500 ea (5 x bags 100ea) | packaging pack of 500 ea | packaging pack of 500 ea (10 x racks 50ea) | packaging case of 36, pack of 6 |
Still not finding the right product?
Explore all of our products under Microtube Centrifuge Tubes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?