Přejít k obsahu
Merck

PCC030C45L

Pellicon® Capsule with Ultracel® Membrane

30 kDa, C Screen, 4.5 m2 manifold with AseptiQuik® L connectors

Synonyma:

Pellicon® Capsule with Ultracel® Membrane

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
41104903
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


Název produktu

Pellicon® Capsule 4.5 m2 Ultracel® 30 kDa C Screen, Feed: AseptiQuik® L Connector Retentate: AseptiQuik® L Connector Permeate: AseptiQuik® L Connector

material

EPDM rubber seal, PPO/PS blend (Core), PPO/PS blend housing, TPE seal, composite regenerated cellulose (CRC) membrane, epoxy adhesive, polyester screen, polypropylene screen, polyurethane adhesive

sterility

sterile; γ-irradiated

product line

Pellicon®

storage condition

room temperature (15-30°C)

parameter

4-8 L/min-m2 flow rate, 50 psi pressure (3.5 bar) at 4-30 °C (Forward Transmembrane Pressure (TMP)), 60 psi max. pressure (4.1 bar) at 4-30 °C (Maximum Inlet Pressure)

technique(s)

ultrafiltration: suitable

H

46.9 cm (18.5 in.)

L

56.2 cm (22.1  in.)

W

48.2 cm (19.0 in.)

filtration area

4.5 m2

screen size

, Type C screen (coarse screen)

volume

1683 mL , in feed channel, 2316 mL , in permeate channel

impurities

≤5 ppm TOC ( in retentate effluent, after a WFI flush of 20 L/m²)

matrix

Ultracel®

pore size

30 kDa

pH range

2-13

fitting

inlet/outlet connection (Feed: AseptiQuik® L Connector), inlet/outlet connection (Permeate: AseptiQuik® L Connector), inlet/outlet connection (Retentate: AseptiQuik® L Connector)

General description

The Pellicon® Capsule is the first of its kind -- a true single-use TFF device that comes ready to process in minutes. Self-contained, it employs an easy-to-use, holderless format and is supplied sterilized by irradiation.

Application

Concentration, Diafiltration, Buffer exchange, Protein purification

Features and Benefits

Plug ′n play, holderless design - easy to install, safe to remove
Gamma Sterilized and preseravtive-free - ready to process in minutes
True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families

Packaging

The manifold assembly is inserted into a polyethylene foam frame and then double bagged and vacuum sealed. The double-bagged assembly is placed within a second foam frame and individually boxed (qty 1 per box). Packaged assemblies are palletized.

Other Notes

The Emprove® Program complements the product portfolio through
Comprehensive documentation to support qualification, risk assessment and process optimization needs.
Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
Convenient 24/7 access to up-to-date product information.

Legal Information

AseptiQuik is a registered trademark of Colder Products Company
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Dodržování zásad SVP pomocí programu Emprove®

Urychlete kvalifikaci, zajistěte shodu s předpisy a ušetřete čas s nástrojem Emprove® Dossiers.

Stáhnout Dossiers

Poznámka: budete přeneseni na stránky EmproveSuite.com. Vaše uživatelské jméno a heslo jsou oddělené od stránek SigmaAldrich.com a mohou se lišit.

Zaregistrujte se zdarma a získejte přístup k Emprove® Dossiers během několika minut. Upgradujte svůj plán předplatného a odemkněte si pokročilý a přizpůsobený obsah v Emprove® Suite.

Nenašli jste, co jste hledali? Navštivte stránku nápovědy programu Emprove®..

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů

Máte potíže při hledání svého čísla produktu nebo šarže/dávky?

Při zobrazení na webových stránkách jsou čísla produktů zkombinována s velikostí balení / množstvím (příklad: T1503-25G). Ujistěte se, že v poli Číslo produktu zadáváte POUZE číslo produktu (příklad: T1503).

Example:

T1503
Číslo produktu
-
25G
Velikost balení / množství

Další příklady:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

zadat jako 1.000309185)

Having trouble? Feel free to contact Technický servis pro pomoc