Merck
NU050/32-221
NovAseptic® Flow Through (1/2") Valve Body 180°
Valve Body - ASTM® A269/270, Electropolished Butt end for welding
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
UNSPSC Code:
23151800
eCl@ss:
32039090
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
Quality Segment
parameter
-1-10 bar max. pressure (-14.5-145 psig) at 25 °C (atmospheric pressure), -80-200 °C max. temp. (-112-392 °F)
General description
NovAseptic® Valves are engineered for optimal performance, reliability, durability and ease of maintenance. NovAseptic® Valves comply with cGMP Design Qualification
criteria for aseptic processing. This is why we choose to call it ‘‘Aseptic by Design.”
Application
NovAseptic® Valves are specifically designed for aseptic applications and comply with the most stringent cleanability and sterilizability requirements. The focus on aseptic design is a significant feature for all valves. Flow through valve is a valve designed for a wide range of applications in the aseptic process; including manifolds, take off and drainage valves.
Biochem/physiol Actions
Net volume (Valve cavity, diaphragm applied): 4mLNet weight (Approximate): Weight kg (lb): 0.1(0.22)Surface roughness: Internal surface Ra ≤ 0.38 μm (15 μin) External surface Ra ≤ 1.6 μm (63 μin)
Other Notes
All open ends on the valve body are protected with plastic covers on delivery and packaged in a closed box.
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)