O této položce
eCl@ss:
37051001
UNSPSC Code:
40141600
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohliQuality Segment
material
plastic handle (Oilon), stainless steel cover (EN 1.4305/ASTM 303), stainless steel indicator (SS 2346)
product line
NovAseptic®
parameter
pressure range at -1-10 bar, temp. range at 0-70 °C
weight
0.4 kg (0.88 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Other Notes
Actuator, Manual
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under NovAseptic® Valve, Manual Plastic Actuator