Přejít k obsahu
Merck

MABF2775

Anti-HCV NS5A Antibody, clone 9E10

Synonyma:

HCV Genome polyprotein;Non-structural Protein 5A

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

NACRES:
NA.43
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
9E10, monoclonal
Application:
ICC, IP, WB
Citations:
-
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

9E10, monoclonal

mol wt

calculated mol wt 327.02 kDa, observed mol wt ~13 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aλ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

vaccinia virus ... ORF45(4961474)

General description

Hepatitis C virus (HCV) is a small (~55-65 nm), enveloped, positive-sense single-stranded RNA virus that is a causative factor for hepatitis C and hepatocellular carcinoma. HCV expresses several proteins that promote viral replication. Genome polyprotein of HCV can be cleaved into 11 different chains. The mature core protein packages viral RNA to form a viral nucleocapsid, and promotes virion budding. It also prevents the establishment of cellular antiviral state by blocking the interferon- a/b and interferon- g signaling pathways and by inducing STAT1 degradation. Envelope proteins E1 and E2 form a heterodimer, which is involved in virus attachment to the host cell, virion internalization through clathrin-dependent endocytosis and fusion with host membrane. Viroprotein P7, a heptameric ion channel protein, plays an essential role in the assembly, envelopment, and secretion of viral particles. HCV polyprotein also contains NS2, a cysteine protease and NS3, a serine protease. NS2 is required for the proteolytic auto-cleavage between NS2 and NS3. NS3 binds a single ATP and catalyzes the unzipping of a single base pair of dsRNA. It also inhibits host antiviral proteins TBK1 and IRF3 and prevents the establishment of an antiviral state. Non-structural protein 4A (NS4A), a peptide cofactor, forms a non-covalent complex with the N-terminal of NS3 and this complex prevents phosphorylation of host IRF3, which also prevents the establishment of dsRNA induced antiviral state. NS4B induces a specific membrane alteration that serves as a scaffold for the virus replication complex. Non-structural protein 5A (NS5A), a multi-functional phosphoprotein, is an RNA-binding peripheral membrane protein that is reported to antagonize numerous cellular pathways, including the antiviral interferon- a response. It can bind to the 3′-ends of HCV plus and minus strand RNAs. NS5A exists as hypo- and hyper-phosphorylated forms and the dynamic transition between these two states is involved in the functions of NS5A. Hyperphosphorylation occurs primarily at its six serine residues within the low complexity sequence of NS5A. Phosphorylated NS5A is shown to be essential for viral replication and assembly. (Ref.: Kandangwa, M. and Liu, Q. (2019). Biochem. Biophys. Res. Commmun. 520(1); 192-197; Saeed, M., et al. (2015). Nature. 524(7566); 471-475; Huang, L., et al. (2005). J. Biol. Chem. 280(43); 36417-36428).

Immunogen

Purified Con1 HCV-non-structural 5A protein.

Application

Quality Control Testing

Evaluated by Western blotting with recombinant Hepatitis C Virus NS5A protein.

Western Blotting Analysis (WB): A 1:100 dilution of this antibody detected recombinant Hepatitis C virus NS5A protein.

Tested Applications

Western Blotting Analysis: A representative lot detected HCV NS5A in Western Blotting applications (Lindenbach, B.D., et al. (2005). Science. 309(5734):623-6).

Immunocytochemistry Analysis: A representative lot detected HCV NS5A in Immunocytochemistry applications (Lindenbach, B.D., et al. (2005). Science. 309(5734):623-6).

Immunoprecipitation Analysis: A representative lot immunoprecipitated HCV NS5A in Immunopreciptation applications (Saeed, M., et al. (2015). Nature. 524(7566):471-5).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-HCV NS5A, clone 9E10, Cat. No. MABF2775, is a mouse monoclonal antibody that detects Hepatitis C virus NS5A protein and is tested for use in Immunocytochemistry, , Immunoprecipitation, and Western Blotting.

Biochem/physiol Actions

Clone 9E10 is a mouse monoclonal antibody that detects non-structural protein 5A (NS5A)of hepatitis C virus.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Vyzkoušejte náš nástroj Nástroj pro výběr produktů a zúžte své možnosti.


Skladovací třída

12 - Non Combustible Liquids

Co se děje?

WGK 1

Bod vzplanutí (°F)

Not applicable

Bod vzplanutí (°C)

Not applicable



Vyberte jednu z posledních verzí:

Osvědčení o analýze (COA)

Lot/Batch Number

Nevidíte správnou verzi?

Potřebujete-li konkrétní verzi, můžete vyhledat daný certifikát podle čísla dávky nebo čísla šarže.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů