Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
UNSPSC Code:
12352203
NACRES:
NA.42
eCl@ss:
32160702
Clone:
MPC-11, monoclonal
Technique(s):
flow cytometry: suitable
Application:
FACS
Citations:
-
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlibiological source
mouse
Quality Segment
antibody form
purified antibody
clone
MPC-11, monoclonal
purified by
affinity chromatography
technique(s)
flow cytometry: suitable
isotype
IgG2b
target post-translational modification
unmodified
General description
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Immunogen
Immunogen information for this antibody is proprietary information.
Application
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Immunoglobulins & Immunology
The isotype control should be used at the same concentration as the primary antibody for which it is a control.
This mouse monoclonal Mouse IgG2b Negative Control, clone MPC-11, FITC conjugate, Cat. No. MABF1783 is validated for use in Flow Cytometry.
Physical form
Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2. This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.
Purified by size-exclusion chromatography
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Skladovací třída
12 - Non Combustible Liquids
wgk
nwg
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.