Přejít k obsahu

HTS189M

ChemiSCREEN Membrane Preparation Recombinant Human GPR103 / QRFP Neuropeptide Receptor

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
41123201
NACRES:
NA.45
eCl@ss:
32161000
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


Quality Segment

description

GPCR Class A, protein target GPR103 / QRFP

technique(s)

radioligand binding assay (RLBA): suitable

UniProt accession no.

Gene Information

human ... QRFPR(84109)

General description

GPR103 is a Gq-coupled receptor that is located in the brain and adrenal gland, and is activated by a 43-amino acid RFamide peptide, QRFP43, and a 26 amino acid fragment, QRFP26/26RFa/Peptide 518 (Jiang et al., 2003; Fukusumi et al., 2003). QRFP43 is the active form in rat brain, and intracerebral administration of QRFP43 induces feeding and metabolic rate (Takayasu et al., 2006; Moriya et al., 2006). Millipore′s GPR103 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of GPR103. The membrane preparations exhibit a Kd of 0.63nM for [125I]-QRFP43. With 10 μg/well GPR103 Membrane Prep and 0.35 nM [125I]- QRFP43, a greater than 6-fold signal-to-background ratio was obtained.
Human GPR103

Application

Radioligand binding assay and GTPγS binding

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Preparation Note

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Analysis Note

Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C

Radioligand: [125I]QRFP43 (Perkin Elmer # NEX408)

Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 6-fold signal:background with 125I-labeled QRFP43 at 0.35 nM.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Vyberte jednu z posledních verzí:

Osvědčení o analýze (COA)

Lot/Batch Number

Nevidíte správnou verzi?

Potřebujete-li konkrétní verzi, můžete vyhledat daný certifikát podle čísla dávky nebo čísla šarže.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů