Přejít k obsahu
Merck

HTS181M

ChemiSCREEN Membrane Preparation Recombinant Human NPBW2 Neuropeptide B/W Receptor

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
41123201
NACRES:
NA.45
eCl@ss:
32161000
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


Quality Segment

description

GPCR Class A, protein target NPBW2 / GPR8

technique(s)

radioligand binding assay (RLBA): suitable

UniProt accession no.

Gene Information

human ... NPBWR2(2832)

General description

FUNCTION: SwissProt: P48146 # Interacts specifically with a number of opioid ligands. Receptor for neuropeptides B and W, which may be involved in neuroendocrine system regulation, food intake and the organization of other signals.
SIZE: 333 amino acids; 36861 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
TISSUE SPECIFICITY: Detected at high levels in caudate nucleus, hyppocampus and amygdala; at moderate levels in the adult brain, thalamus, parietal cortex, pituitary gland, adrenal gland and lymph nodes.
SIMILARITY: SwissProt: P48146 ## Belongs to the G-protein coupled receptor 1 family.

Application

Radioligand binding assay and GTPγS binding

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80oC.

Preparation Note

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Analysis Note

Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM Tris, pH 7.4. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, filtered and stored at 4°C.
Radioligand: [125I]-neuropeptide W23 (Perkin Elmer#: NET-403)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl, filtered and stored at 4°C.

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 10-fold signal:background with 125I-labeled neuropeptide W23 at 0.35nM.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů