Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
UNSPSC Code:
41106514
NACRES:
NA.84
eCl@ss:
32161000
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlibiological source
human
Quality Segment
recombinant
expressed in Chem-1 cells
manufacturer/tradename
ChemiScreen, Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS), radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
-65 to -96°C
General description
Full-length human GABBR1b and GPR51 cDNAs encoding GABAB1b/GABAB2
The neurotransmitter γ-aminobutyric acid (GABA) exerts its effects through an ion channel, GABAA, and a GPCR, GABAB. Functional GABAB is a heterodimer composed of the GABAB1 and GABAB2 subunits, which share 35% sequence identity and belong to the class 3 family of GPCRs. The GABAB1 subunit, which exists as splice variants GABAB1a and GABAB1b, binds directly to GABA and is required for agonist activation. The GABAB2 and GABAB1 subunits associate by formation of a coiled coil by their C-terminal tails; this association masks an ER retention sequence in GABAB1 to permit export from the ER and trafficking to the cell surface. In addition to its chaperone function, GABAB2 is the component that couples to Gi to reduce intracellular cAMP. Agonists of GABAB, such as baclofen, are used clinically for treatment of muscle spasticity, migraine headache and musculoskeletal pain (Bowery et al., 2002). Chemicon′s GABAB1b membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of GABAB1b receptor interactions with its ligand. The membrane preparations exhibit a Kd of 1.15nM for [3H]-CGP 54626A. With 5.0 ug/well GABAB1b Membrane Prep and 7.5nM [3H]-CGP 54626A, a greater than 5-fold signal-to-background ratio was obtained.
Application
Radioligand binding assay and GTPγS binding
Biochem/physiol Actions
GPCR Class: C
Protein Target: GABAB1b
Target Sub-Family: GABAB
Features and Benefits
Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM Tris, pH 7.4. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 1X Hanks′ Balanced Salt Solution (HBSS): GIBCO Cat. No. 14175
Radioligand: [3H]-CGP 54626A (American Radiolabeled Chemicals Inc., ART 517)
Wash Buffer: 1X Hanks′ Balanced Salt Solution (HBSS): GIBCO Cat. No. 14175
One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 5-fold signal:background with 3H-labeled CGP 54626A at 7.5nM.
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM Tris, pH 7.4. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 1X Hanks′ Balanced Salt Solution (HBSS): GIBCO Cat. No. 14175
Radioligand: [3H]-CGP 54626A (American Radiolabeled Chemicals Inc., ART 517)
Wash Buffer: 1X Hanks′ Balanced Salt Solution (HBSS): GIBCO Cat. No. 14175
One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 5-fold signal:background with 3H-labeled CGP 54626A at 7.5nM.
Physical form
Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Packaging method: Membranes protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Preparation Note
Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.
Analysis Note
Table 1. Signal:background and specific binding values obtained in a competition binding assay with 5ug/well of GABAB1b Receptor membrane prep.
SPECIFICATIONS: 1 unit = 5.0 µg membrane preparation
Bmax: 8.49 pmol/mg
Kd: 1.15 nM
| 5 µg/well | |
|---|---|
| Signal:Background | 6.4 |
| Specific Binding (cpm) | 2879 |
SPECIFICATIONS: 1 unit = 5.0 µg membrane preparation
Bmax: 8.49 pmol/mg
Kd: 1.15 nM
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Skladovací třída
12 - Non Combustible Liquids
Co se děje?
WGK 2
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.