Přejít k obsahu
Merck

HTS025M

ChemiSCREEN Membrane Preparation Recombinant Human MCH1 Receptor

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
41123201
NACRES:
NA.45
eCl@ss:
32161000
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


Quality Segment

description

GPCR Class A, protein target MCHR1 / GPR24

technique(s)

radioligand binding assay (RLBA): suitable

UniProt accession no.

Gene Information

human ... MCHR1(2847)

General description

Full length human MCHR1 cDNA encoding MCH1
Melanin-concentrating hormone (MCH) is a cyclic, 19 amino acid peptide produced in the lateral hypothalamus and other regions of the mammalian brain (Bittencourt et al., 1992). When administered intracerebrally or overexpressed transgenically in rodents, MCH stimulates food intake and weight gain (Ludwig et al., 2001). A receptor for MCH, previously identified as an orphan receptor SLC-1 and renamed MCH1, has been identified in humans and rodents. Consistent with a role in hypothalmic control of feeding behavior, MCH1 is expressed in in the ventromedial and dorsomedial nuclei of the hypothalamus (Chambers et al., 1999). Genetic ablation and pharmacological antagonism of MCH1 in rodents results in resistance to obesity caused by a high fat diet or leptin deficiency (Borowsky et al., 2002; Segal-Lieberman, et al., 2003; Shearman et al., 2003; Marsh et al., 2002; Chen et al., 2002). Thus, MCH1 is an attractive target for obesity. Millipore′s MCH1membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level GPCR surface expression; thus, they are ideal HTS tools for screening for agonists and antagonists of MCH1. The membrane preparations exhibit a Kd of 0.41 nM for [125I]-MCH. With 0.4 nM for [125I]-MCH, 5 μg/well MCH1 Membrane Prep yields greater than 8 fold signal-to-background ratio.

Application

Radioligand binding assay and GTPγS binding

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 8-fold signal:background with 125I-labeled MCH at 0.4 nM.

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Preparation Note

Store at –70°C. Product is stable for at least 6 months from the date of receipt when stored as directed. Do not freeze and thaw.

Analysis Note

RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C

Radioligand: [125I]-MCH (Perkin Elmer NEX373)

Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů