Hlavní dokumenty
G30-011
NovAseptic® Mixer
About This Item
UNSPSC Code:
41103801
eCl@ss:
32010600
Informace o stanovených cenách a dostupnosti nejsou v současnosti dostupné.
material
316L stainless steel bar ((ASME SA-479)/EN 1.4435 (EN 10272))
product line
NovAseptic®
parameter
pressure range at -1-10 bar
temp. range at -80-200 °C
diam.
160 mm (6.30 in.) , welding
weight
2.5 kg (5.51 lb)
Application
The assembled NovAseptic® Mixer may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Other Notes
Surface Finish, Exterior: Liquid contact surface, manually polished; Ra ≤ 0.5µm (20µin)
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
The NOVASEPTIC® Mixer is classified in Europe, for PED Directive 97/23/EC Category IV and approved by Notified Body. The Tank Plate is designed to meet requirements for USA, ASME VIII Div 2. Stresses in Tank Plate due to internal pressure have been analyse