Přejít k obsahu
Merck

ABN1342

Anti-Septin 11 Antibody

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


conjugate

unconjugated

Quality Segment

antibody form

serum

clone

polyclonal

purified by

(unpurified)

species reactivity

rat, rabbit

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

KLH-conjugated linear peptide corresponding to the N-terminal sequence of rat Septin 11.
~51 kDa observed. This antibody is predicted to react with all isoforms. Uncharacterized band(s) may appear in some lysates.

Application

Epitope: N-terminus

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.

Preparation Note

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: A 1:2,500 dilution of this antibody detected Septin11 in 10 µg of rat brain tissue lysate.

Other Notes

Rat. Predicted to react with Mouse, Human, Horse, Porcine based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů