Key Documents
2711-10100
Millipore
NovaSeptum® Cell Culture sampling Unit
High Purity Application; 100 mL
About This Item
UNSPSC Code:
41104105
eCl@ss:
36101790
material
316 stainless steel cannula (ASTM)
polyester body
septum (Medical grade platinum-cured silicone)
Quality Level
product line
NovaSeptum®
manufacturer/tradename
NovaSeptum®
Application
Bacterial counting and sampling from bioreactors
Legal Information
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany