O této položce
NACRES:
NB.51
UNSPSC Code:
41121804
Capacity:
250 mL, 250 mL
Material:
borosilicate glass flask, pear-shaped , single-neck
Joint:
joint: ST/NS24/40
Packaging:
pkg of 1 ea
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
borosilicate glass flask, pear-shaped , single-neck
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F314250
capacity
250 mL, 250 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Explore all of our products under Synthware™ recovery flask
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?