Přejít k obsahu
MT51343151
Mettler Toledo InLab® Viscous & Viscous Pro pH electrode
Temperature probe NTC 30 kOhm
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
NACRES:
NB.79
UNSPSC Code:
41115604
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
glass membrane (polymer), glass shaft (HA)
description
MultiPin connector, Without cable
manufacturer/tradename
Mettler-Toledo 51343151
L × W
40 mm × 6 mm
General description
The InLab® Viscous has been especially developed for viscous and highly glutinous samples.
The SteadyForce reference system guarantees the discharge of electrolytes, even with the most glutinous and fatty of samples, e.g. cosmetics, paints or resins.
The form has been chosen so that as little of the sample as possible adheres to the shaft and to facilitate cleaning. The result is a problem solver for most difficult sample types, the highly viscous materials.
  • high performance puncture electrode - especially designed for highly viscous
  • samples
  • easy to clean
  • SteadyForce reference system
Legal Information
InLab is a registered trademark of Mettler-Toledo, Inc.
MultiPin is a trademark of Mettler-Toledo, Inc.
SteadyForce is a trademark of Mettler-Toledo, Inc.
Still not finding the right product?
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)