Merck
BMSH6002EUK
Incu-Mixer MP2 Heated 2 Plate Vortexer 115-240 V
AC/DC input 115 V / 240 V AC, UK plug
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
NACRES:
NB.77
UNSPSC Code:
41103800
eCl@ss:
32039090
Manufacturer/tradename:
Benchmark Scientific H6002-E-UK
Parameter:
100 rpm speed (1200 rpm)
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
manufacturer/tradename
Benchmark Scientific H6002-E-UK
parameter
100 rpm speed (1200 rpm)
W × D × H
11 in. × 10.6 in. × 5.5 in.
size
, no of position=4
General description
The new Incu-Mixer MP series is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally, the heated lid minimizes evaporation and prevents condensation.
A compact, 2mm orbit ensures thorough mixing in microplates without splashing. Speed, time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy.
Designed for application flexibility, the Incu-Mixer series is available in two, and four-plate capacities, both capable of accepting all standard plates up to 40mm high. Independent time, temperature and speed controls allow the mixer to function as 3 machines in one; a multi-plate incubator, vortexer, or a combination of the two. The Incu-Mixer MP series is ideal for a wide variety of molecular biology applications including immunochemical reactions, enzyme and protein analysis, and microarray analysis.
  • Optimized for incubated mixing in microplates
  • Heated lid prevents condensation
  • Independently control time, temp. and speed
Still not finding the right product?
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)