Merck
  • Metabolism and mis-metabolism of the neuropathological signature protein TDP-43.
Metabolism and mis-metabolism of the neuropathological signature protein TDP-43.
Journal of cell science (2014-05-27)
Chi-Chen Huang, Jayarama Krishnan Bose, Pritha Majumder, Kuen-Haur Lee, Jen-Tse Joseph Huang, Jeffrey K Huang, Che-Kun James Shen
ABSTRACT
TDP-43 (also known as TARDBP) is a pathological signature protein of neurodegenerative diseases, with TDP-43 proteinopathies including frontotemporal lobar degeneration (FTLD)-TDP and amyotrophic lateral sclerosis (ALS)-TDP. These TDP-43 proteinopathies are characterized by cytoplasmic insoluble TDP-43-positive aggregates in the diseased cells, the formation of which requires the seeding of TDP-25 fragment generated by caspase cleavage of TDP-43. We have investigated the metabolism and mis-metabolism of TDP-43 in cultured cells and found that endogenous and exogenously overexpressed TDP-43 is degraded not only by the ubiquitin proteasome system (UPS) and macroautophagy, but also by the chaperone-mediated autophagy (CMA) mediated through an interaction between Hsc70 (also known as HSPA8) and ubiquitylated TDP-43. Furthermore, proteolytic cleavage of TDP-43 by caspase(s) is a necessary intermediate step for degradation of the majority of the TDP-43 protein, with the TDP-25 and TDP-35 fragments being the main substrates. Finally, we have determined the threshold level of the TDP-25 fragment that is necessary for formation of the cytosolic TDP-43-positive aggregates in cells containing the full-length TDP-43 at an elevated level close to that found in patients with TDP-43 proteinopathies. A comprehensive model of the metabolism and mis-metabolism of TDP-43 in relation to these findings is presented.
Sigma-Aldrich
Anti-TARDBP (AB1) antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-GTF2H1 antibody produced in rabbit, IgG fraction of antiserum
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology