Merck
  • The use of tenoten and tenoten (pediatric formulation) as a drug for premedication in adults and children during outpatients dentist visit.
The use of tenoten and tenoten (pediatric formulation) as a drug for premedication in adults and children during outpatients dentist visit.
Bulletin of experimental biology and medicine (2009-12-23)
L I Larentsova, D V Lyubimova, T Yu Fokina, E A Rvacheva, D V Sosul'nikov
ABSTRACT
The use of tenoten and tenoten (pediatric formulation) for premedication in adults and children before dental treatment reduces patient anxiety; the effect is comparable with that of classical anxiolytics and is not accompanied by side effects. These preparations can be recommended for wide use in dentistry.
Product Description
Supelco
Phenazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®