Merck
  • Human Autosomal Recessive DNA Polymerase Delta 3 Deficiency Presenting as Omenn Syndrome.
Human Autosomal Recessive DNA Polymerase Delta 3 Deficiency Presenting as Omenn Syndrome.
Journal of clinical immunology (2023-12-15)
Maria Rodrigo Riestra, Bethany A Pillay, Mathijs Willemsen, Verena Kienapfel, Lisa Ehlers, Selket Delafontaine, Antoine Pinton, Marjon Wouters, Anneleen Hombrouck, Kate Sauer, Xavier Bossuyt, Arnout Voet, Stefaan J Soenen, Cecilia Dominguez Conde, Giorgia Bucciol, Kaan Boztug, Stephanie Humblet-Baron, Aurore Touzart, Frédéric Rieux-Laucat, Luigi D Notarangelo, Leen Moens, Isabelle Meyts
ABSTRACT
The DNA polymerase δ complex (PolD), comprising catalytic subunit POLD1 and accessory subunits POLD2, POLD3, and POLD4, is essential for DNA synthesis and is central to genome integrity. We identified, by whole exome sequencing, a homozygous missense mutation (c.1118A > C; p.K373T) in POLD3 in a patient with Omenn syndrome. The patient exhibited severely decreased numbers of naïve T cells associated with a restricted T-cell receptor repertoire and a defect in the early stages of TCR recombination. The patient received hematopoietic stem cell transplantation at age 6 months. He manifested progressive neurological regression and ultimately died at age 4 years. We performed molecular and functional analysis of the mutant POLD3 and assessed cell cycle progression as well as replication-associated DNA damage. Patient fibroblasts showed a marked defect in S-phase entry and an enhanced number of double-stranded DNA break-associated foci despite normal expression levels of PolD components. The cell cycle defect was rescued by transduction with WT POLD3. This study validates autosomal recessive POLD3 deficiency as a novel cause of profound T-cell deficiency and Omenn syndrome.
Sigma-Aldrich
Anti-POLD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse