Merck
SOAT1 promotes mevalonate pathway dependency in pancreatic cancer.
The Journal of experimental medicine (2020-07-08)
Tobiloba E Oni, Giulia Biffi, Lindsey A Baker, Yuan Hao, Claudia Tonelli, Tim D D Somerville, Astrid Deschênes, Pascal Belleau, Chang-Il Hwang, Francisco J Sánchez-Rivera, Hilary Cox, Erin Brosnan, Abhishek Doshi, Rebecca P Lumia, Kimia Khaledi, Youngkyu Park, Lloyd C Trotman, Scott W Lowe, Alexander Krasnitz, Christopher R Vakoc, David A Tuveson
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) has a dismal prognosis, and new therapies are needed. Altered metabolism is a cancer vulnerability, and several metabolic pathways have been shown to promote PDAC. However, the changes in cholesterol metabolism and their role during PDAC progression remain largely unknown. Here we used organoid and mouse models to determine the drivers of altered cholesterol metabolism in PDAC and the consequences of its disruption on tumor progression. We identified sterol O-acyltransferase 1 (SOAT1) as a key player in sustaining the mevalonate pathway by converting cholesterol to inert cholesterol esters, thereby preventing the negative feedback elicited by unesterified cholesterol. Genetic targeting of Soat1 impairs cell proliferation in vitro and tumor progression in vivo and reveals a mevalonate pathway dependency in p53 mutant PDAC cells that have undergone p53 loss of heterozygosity (LOH). In contrast, pancreatic organoids lacking p53 mutation and p53 LOH are insensitive to SOAT1 loss, indicating a potential therapeutic window for inhibiting SOAT1 in PDAC.
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Farnesyl pyrophosphate ammonium salt, methanol:ammonia solution, ≥95% (TLC)
Sigma-Aldrich
Cholesterol-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Geranylgeranyl pyrophosphate ammonium salt, solution, ≥95% (TLC), ~1 mg/mL in methanol: NH4OH (7:3)
Sigma-Aldrich
Anti-Heat Shock Protein 90α Antibody, from rabbit