Merck
  • Superoxide anions modulate the effects of alarin in the paraventricular nucleus on sympathetic activity and blood pressure in spontaneously hypertensive rats.
Superoxide anions modulate the effects of alarin in the paraventricular nucleus on sympathetic activity and blood pressure in spontaneously hypertensive rats.
Neuropeptides (2020-02-09)
Qian Wang, Fanxin Deng, Dawei Zhu
ABSTRACT
Neuropeptides are involved in the regulation of the sympathetic activity and blood pressure in the paraventricular nucleus of the hypothalamus (PVN). The present study was designed to determine how alarin modulates the renal sympathetic nerve activity (RSNA), arterial blood pressure and mean arterial pressure (MAP) in the PVN, and whether superoxide anions regulate the effects of alarin in the PVN of spontaneously hypertensive rats (SHRs). Acute experiment was carried out with male Wistar-Kyoto rats (WKY) and SHRs under anesthesia. RSNA, systolic blood pressure (SBP), diastolic blood pressure (DBP), and MAP were measured. Alarin microinjection into the PVN increased RSNA (7.8 ± 1.8 vs. 14.8 ± 2.3%), SBP (5.9 ± 1.4 vs. 12.1 ± 1.6 mmHg), DBP (5.1 ± 0.8 vs. 10.0 ± 1.1 mmHg), and MAP (5.4 ± 1.2 vs. 10.7 ± 1.3 mmHg) in WKY rats and SHRs,. Alarin antagonist ala6-25 Cys decreased RSNA, SBP, DBP, and MAP in SHRs, and inhibited the effects of alarin. The alarin level was increased in the PVN of SHR compared to WKY rats. (29.7 ± 4.9 vs. 14.6 ± 2.4 pg/mg protein). PVN microinjection of superoxide anion scavengers tempol and tiron, or NAD(P)H oxidase inhibitor apocynin, decreased RSNA, SBP, DBP, and MAP in SHRs, and inhibited the effects of alarin, but the superoxide dismutase inhibitor diethyldithiocarbamic acid potentiated the effects of alarin. Superoxide anions and NAD(P)H oxidase activity levels in the PVN were increased by alarin, but decreased by alarin antagonist ala6-25 Cys. The alarin-induced increases in superoxide anions and NAD(P)H oxidase activity levels were abolished by pre-treatment with ala6-25 Cys. The results suggest that alarin in the PVN increases sympathetic outflow and blood pressure. The enhanced activity of endogenous alarin in the PVN contributes to sympathetic activation in hypertension, and the superoxide anion is involved in these alarin-mediated processes in the PVN.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Diethyldithiocarbamic acid diethylammonium salt, 97%
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidinol, 98%