Nitrilotriacetic acid
Synonym(s):
N,N-Bis(carboxymethyl)glycine, NTA, Tris(carboxymethyl)amine, Nitrilotriacetic acid
Linear Formula:
N(CH2COOH)3
CAS Number:
Molecular Weight:
191.14
Products
Product Category
Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reagent Type
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing