504327
PTFE Frit, 20 μm porosity
for use with 6mL glass SPE tubes, pk of 60
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
material
PTFE frit
description
for SPE Tube
packaging
pk of 60
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with 6mL glass SPE tubes
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Porous frits are fitted at the column′s outlet end. This helps to retain the support material in the column. It could also be fitted in the coupling holding the column to the detector, this helps unpacking the column or even changing the frit on blockage. Frit porosity depends on the particles of the smallest diameter in the support material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
N.A. Parris
Instrumental Liquid Chromatography: A Practical Manual on High-Performance Liquid Chromatographic Methods, 48-48 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service