Skip to Content
Merck
All Photos(1)

Key Documents

Z743345

Laser Cryo-Clear® labels

for 1.5 mL tubes, L × W 1.28 in. × 0.5 in., pkg of 1700 pkg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103032
NACRES:
NB.35

Pricing and availability is not currently available.

packaging

pkg of 1700 pkg

manufacturer/tradename

Diversified Biotech CLEAR-170

technique(s)

cryopreservation: suitable

L × W

1.28 in. × 0.5 in.

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
Z743344Z743417Z743405
manufacturer/tradename

Diversified Biotech CLEAR-170

manufacturer/tradename

Diversified Biotech CLEAR-238

manufacturer/tradename

Diversified Biotech LCRY-1300-G

manufacturer/tradename

Diversified Biotech LCRY-1000-R

packaging

pkg of 1700 pkg

packaging

pkg of 2380 pkg

packaging

pkg of 840 labels 20 sheets of 42 labels

packaging

pkg of 20 labels 20 sheets of 1 label

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable

technique(s)

cryopreservation: suitable

General description

Laser Cryo-Clear® labels

Legal Information

Cryo-Clear is a registered trademark of Diversified Biotech

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service