Z654442
Consort EV series power supplies
EV2650, output0-600 V, 0-500 mA, 0-150 W
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
AC/DC input
230 V AC
description
30 ohms min load resistance
4 × 4 mm sockets
manufacturer/tradename
EV2650
availability
available only in EU
parameter
0-40 °C ambient temperature
W × D × H
24 cm × 20 cm × 13 cm
weight
4.3 kg
output
0-600 V, 0-500 mA, 0-150 W
Looking for similar products? Visit Product Comparison Guide
General description
- Timer 0-99.59 hour
- Volt hours 0-99.99 kVH
- LCD display, 2 × 16 characters
- Ground leakage detection 500 μA
- No load detection, programmable, overload detection
- 3600 values of data logging
- 0-95% non condensing humidity
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service