WPB01451WA
Whirl-Pak® Stand-Up Bag
2041 mL capacity(69 oz)
Synonym(s):
Stand-Up Bag, Stand-Up Storage Bag, Whirl-Pak Bag
About This Item
Recommended Products
sterility
sterile
packaging
pkg of 250 ea
manufacturer/tradename
B01451WA (Nasco)
parameter
-346-180 °F temp. range (-210 to 82 °C)
W × L × thickness
19 cm × 38 cm × 0.102 mm
capacity
2041 mL × 69 oz
2041 mL capacity(69 oz)
Related Categories
General description
The white, write-on strip makes sample identification easier and more convenient. It′s easy to write on this white strip using an ordinary ballpoint pen, pencil, or felt-tip marker. The paper-like finish virtually eliminates ink smearing, and provides a surface that will accept a variety of writing instruments.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)