TMO267544
Nunc® Edge 2.0 96-well Plate
untreated, polystyrene, sterile, with lid, clear, case of 160 ea
Synonym(s):
cell culture plate, cell growth plate
All Photos(1)
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
Recommended Products
material
clear
polystyrene
sterility
sterile
packaging
case of 160 ea
manufacturer/tradename
Nunc 267544
Related Categories
General description
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)