Skip to Content
Merck
All Photos(1)

Key Documents

T4202

Sigma-Aldrich

TRKA (440-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp781l14186, MTC, NTRK1, TRK1, p140-TrKA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

19-25 nmol/min·mg

mol wt

~66 kDa

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NTRK1(4914)

Application

TRKA (440-end), active, GST tagged human was used to study functional NGF signal transduction.

Biochem/physiol Actions

TRKA (440-end), active, GST tagged human is a 140 kD membrane-spanning protein tyrosine kinase that belongs to proto-oncogene family. TRKA facilitates the regulation of cell differentiation and survival via activating SHC1 and FRS2 which in turn activates a GRB2-Ras-MAPK cascade. It also plays a pivotal role in controlling NF-κ-B activation as well as transcription of genes involved in cell survival via PLCG1.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Iannone et al.
Rheumatology (Oxford, England), 41(12), 1413-1418 (2002-12-07)
We aimed to investigate the expression of nerve growth factor (NGF) and high affinity NGF receptor (p140 TrkA) on chondrocytes from human healthy and osteoarthritic cartilage. We recruited 12 patients with osteoarthritis (OA) undergoing surgical knee replacement. Articular cartilage was
Ilaria Caon et al.
Cancers, 13(5) (2021-04-04)
Interaction between cancer cells and their microenvironment is central in defining the fate of cancer development. Tumour cells secrete signals (cytokines, chemokines, growth factors) that modify the surrounding area, while the niche supplies structures and activities necessary for tumour maintenance
F Boissière et al.
Molecular and chemical neuropathology, 28(1-3), 219-223 (1996-05-01)
The most consistent neuropathological finding in Alzheimer disease (AD) is the loss of cholinergic neurons of the nucleus basalis of Meynert (NbM). Using immunohistochemistry, we have previously shown that cholinergic neurons located in the ventral striatum were affected, whereas those
R M Stephens et al.
Neuron, 12(3), 691-705 (1994-03-01)
In response to NGF, the Trk receptor tyrosine kinase forms a complex with SHC, a protein that couples receptor tyrosine kinases to p21ras. Complex formation between Trk and SHC, SHC tyrosine phosphorylation, and association of SHC with Grb2 were mediated
D R Kaplan et al.
Science (New York, N.Y.), 252(5005), 554-558 (1991-04-26)
The trk proto-oncogene encodes a 140-kilodalton, membrane-spanning protein tyrosine kinase (p140prototrk) that is expressed only in neural tissues. Nerve growth factor (NGF) stimulates phosphorylation of p140prototrk in neural cell lines and in embryonic dorsal root ganglia. Affinity cross-linking and equilibrium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service