Skip to Content
Merck
All Photos(1)

Key Documents

SRP8052

Sigma-Aldrich

CD279 (human): FC (human)

recombinant, expressed in CHO cells, ≥98% (SDS-PAGE)

Synonym(s):

Hepatitis A virus cellular receptor 2, PD-1, Programmed cell death protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€134.70
250 MG
€173.70

Vendor SKU: EN300-228616-100MG

€134.70


Availability
Ships in 14 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
100 MG
€134.70
250 MG
€173.70

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

Vendor SKU: EN300-228616-100MG

€134.70


Availability
Ships in 14 days - from Aldrich Partner.

To order products, please contact your local dealer.

biological source

human

recombinant

expressed in CHO cells

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

monomer 46 kDa by calculation

packaging

pkg of 100 μg

impurities

<0.06 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

General description

Programmed Death-1 (PD-1, CD279) is a type I transmembrane protein belonging to the CD28/CTLA-4 family of immunoreceptors that mediate signals for regulating immune responses. Members of the CD28/CTLA-4 family have been shown to either promote T cell activation (CD28 and ICOS) or downregulate T cell activation (CTLA-4 and PD-1). CD279 is expressed on activated T-cells, B-cells, myeloid cells and on a subset of thymocytes. In vitro, ligation of CD279 inhibits TCR-mediated T-cell proliferation and production of IL-1, IL-4, IL-10 and IFN-γ. In addition, CD279 ligation also inhibits BCR mediated signaling. CD279 deficient mice have a defect in peripheral tolerance and spontaneously develop autoimmune diseases.

Physical form

Lyophilized from 0.2 μm-filtered solution in PBS.

Reconstitution

Reconstitute at 100 μg/mL in sterile PBS.

Other Notes

The extracellular domain of human CD279 [PD-1] (aa 25-167) is fused to the N-terminus of the Fc region of human IgG1.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service