Skip to Content
Merck
All Photos(2)

Key Documents

SRP5169

Sigma-Aldrich

Calmodulin1, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CALM1, CALML2, CAMI, DD132, PHKD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~17 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CALM1(801)

General description

Calmodulin 1 is a member of calcium-modulated proteins which is present in the cytosol and on membranes facing the cytosol and has a high affinity for calcium. Calmodulin 1 has 4 calcium-binding domains and plays a role in cell growth, cell cycle, signal transduction and the synthesis and release of neurotransmitters. Calmodulin can bind to the epidermal growth factor receptor at its cytosolic juxtamembrane region and this inhibits its tyrosine kinase activity. A number of other proteins including HSP70 have been shown to interact with Calmodulin 1 in a cell-phase-specific manner.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Huang et al.
Cell stress & chaperones, 14(4), 343-353 (2008-11-08)
The cell cycle is controlled by regulators functioning at the right time and at the right place. We have found that calmodulin (CaM) has specific distribution patterns during different cell-cycle stages. Here, we identify cell-cycle-specific binding proteins of CaM and
Hongbing Li et al.
FEBS letters, 559(1-3), 175-180 (2004-02-13)
We have previously shown that exogenous calmodulin (CaM) binds to the epidermal growth factor receptor (EGFR) at its cytosolic juxtamembrane region inhibiting its tyrosine kinase activity. We demonstrate in this report that endogenous CaM binds to EGFR in intact cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service