Skip to Content
Merck
All Photos(2)

Key Documents

SML3931

Sigma-Aldrich

(-)-Vestitol

≥98% (HPLC)

Synonym(s):

(3R)-3,4-Dihydro-3-(2-hydroxy-4-methoxyphenyl)-2H-1-benzopyran-7-ol, (3R)-Vestitol, Vestitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O4
CAS Number:
Molecular Weight:
272.30
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Hydroxyisoflavan isolated from Cuban and Brazilian red propolis that exhibits potent anti-inflammatory, anti-viral, and antiparasitic activities.


Vestitol is a hydroxyisoflavan isolated from Cuban and Brazilian red propolis, licorice root and other sources that exhibits potent anti-inflammatory, anti-viral, and antiparasitic activities. Apparently Vestitol inhibits release of the chemokines CXCL1/KC and CXCL2/MIP-2 from macrophages. Vestitol strongly inhibits Chikungunya virus infection. Vestitol exhibits anti-trypanosomal activity against T. brucei and T. congolense (EC50 3.86 and 4.36 µg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Activity of Red Nigerian Propolis and Some of Its Components against Trypanosoma brucei and Trypanosoma congolense
Molecules (Basel), 28(2), 622-622 (2023)
Potential in vitro anti-periodontopathogenic, anti-Chikungunya activities and in vivo toxicity of Brazilian red propolis
Scientific Reports, 12(1), 21165-21165 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service