Skip to Content
Merck
All Photos(1)

Key Documents

SML1488

Sigma-Aldrich

Metamizole sodium hydrate

≥98% (HPLC)

Synonym(s):

(antipyrinylmethylamino)methanesulfonic acid sodium salt, 1-Phenyl-2,3-dimethyl-5-pyrazolone-4-methylaminomethanesulfonate sodium, 4-Methylamino-1,5-dimethyl-2-phenyl-3-pyrazolone sodium methanesulfonate, Analgin, Dipyrone, Metamizol, Methampyrone, Methylmelubrin, Noraminopyrine methanesulfonate sodium, Sodium methylaminoantipyrine methanesulfonate, Sulpyrine, [(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]methanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N3O4S · Na · H2O
CAS Number:
Molecular Weight:
351.35
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

[Na+].[S](=O)(=O)([O-])CN(C)c1[c]([n]([n](c1C)C)c2ccccc2)=O.O

InChI

1S/C13H17N3O4S.Na.H2O/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;;/h4-8H,9H2,1-3H3,(H,18,19,20);;1H2/q;+1;/p-1

InChI key

UNZIDPIPYUMVPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Metamizole sodium is a nonsteroidal anti-inflammatory drug (NSAID). It is widely used as an analgesic and antipyretic agent.[1]

Biochem/physiol Actions

Metamizole (Dipyrone) is a potent analgesic and antipyretic.
Metamizole (Dipyrone) is a potent analgesic and antipyretic. Metamizole inhibits prostaglandin synthesis in both the periphery and the central nervous system. Metamizole may act as a prodrug. It is rapidly hydrolyzed to N-methyl-4-aminoantipyrine (MAA) and 4-aminoantipyrine (AA) which inhibit cyclo-oxygenase activity and can be further acylated with arachidonic acid forming compounds that bind to central and peripheral cannabinoid (CB1 and CB2) receptors and TRPV1 receptors. It is believed that these metabolites play a major role in metamizole activity. Metamizole has been withdrawn from clinical use in several countries because of a side-effect of agranulocytosis in some patients.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acute kidney injury associated with metamizole sodium ingestion.
Hassan K, et al.
Renal Failure, 33(5), 544-547 (2011)
Diéssica Padilha Dalenogare et al.
Pharmaceuticals (Basel, Switzerland), 14(8) (2021-08-29)
Headaches are frequently described in progressive multiple sclerosis (PMS) patients, but their mechanism remains unknown. Transient receptor potential ankyrin 1 (TRPA1) was involved in neuropathic nociception in a model of PMS induced by experimental autoimmune encephalomyelitis (PMS-EAE), and TRPA1 activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service