Skip to Content
Merck
All Photos(2)

Key Documents

SAB5600064

Sigma-Aldrich

Anti-Phospho-akt (ser473) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM251, purified immunoglobulin

Synonym(s):

phosphorylation at Ser473

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM251, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 1:1000-1:2000
immunohistochemistry: 1:200-1:500

isotype

IgG

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer473)

Gene Information

human ... AKT1(207)

General description

AKT1 (AKT serine/threonine kinase 1) is also called as v-akt murine thymoma viral oncogene homolog 1 and PKB α (protein kinase B). It is expressed in liver,muscle and adipocytes. AKT1 gene is mapped to human chromosome 14q32. It is a member of AKT family that has an amino-terminal pleckstrin homology (PH) domain, a short α helical linker and a carboxyl-terminal kinase domain.

Specificity

This antibody reacts to Akt only when phosphorylated at Ser473. There is no cross-reactivity with Akt without phosphorylation at Ser473. This antibody may also react to bovine, mouse or rat Phospho-Akt (Ser473), as predicted by immunogen homology

Immunogen

Synthetic peptide corresponding to human Phospho-Akt (Ser473)

Biochem/physiol Actions

AKT1 (AKT serine/threonine kinase 1) controls the survival of cells and anti-apoptotic actions that attack the pathogenesis of several cancers, hence it plays a crucial role in tumorigenesis. In mice, AKT1 is essential for normal development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association between single nucleotide polymorphisms in AKT1 and the risk of prostate cancer in the Chinese Han population.
Liu JM, et al.
Genetics and molecular research : GMR, 16(1) (2017)
Claudia Delgadillo-Puga et al.
Nutrients, 15(11) (2023-06-10)
Pecans (Carya illinoinensis) are considered a functional food due to the high content of polyunsaturated fatty acids, dietary fiber and polyphenols. To determine the effect of whole pecans (WP) or a pecan polyphenol (PP) extract on the development of metabolic
AKT plays a central role in tumorigenesis
Testa JR and Bellacosa A
Proceedings of the National Academy of Sciences of the USA, 98(20), 10983-10985 (2001)
Eun-Hui Lee et al.
Scientific reports, 8(1), 9894-9894 (2018-07-04)
Accumulating data have indicated a fundamental role of eosinophils in regulating adipose tissue homeostasis. Here, we performed whole-genome RNA sequencing of the small intestinal tract, which suggested the presence of impaired lipid metabolism in eosinophil-deficient ΔdblGATA mice. ΔdblGATA mice fed
Akt1/PKBalpha is required for normal growth but dispensable for maintenance of glucose homeostasis in mice
Cho H, et al.
The Journal of Biological Chemistry, 276(42), 38349-38352 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service