Skip to Content
Merck
All Photos(2)

Key Documents

SAB2105467

Sigma-Aldrich

Anti-KIAA1199 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-TMEM2L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

153 kDa

species reactivity

human, dog, rat, bovine, horse, mouse, guinea pig, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

Immunogen

Synthetic peptide directed towards the middle region of human KIAA1199

Application

Anti-KIAA1199 antibody produced in rabbit is suitable for western blot.

Sequence

Synthetic peptide located within the following region: PFLSIISARYSPHQDADPLKPREPAIIRHFIAYKNQDHGAWLRGGDVWLD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Shiozawa et al.
Scientific reports, 12(1), 17242-17242 (2022-10-15)
Destruction of articular cartilage in osteoarthritis (OA) is initiated by depletion of the hyaluronan (HA)-aggrecan network, followed by degradation of the collagen fibrils. Previously, we reported the implications of HA-binding protein involved in HA depolymerization (HYBID), alias cell migration-inducing protein
Takashi Ohtsuki et al.
International journal of molecular sciences, 21(9) (2020-05-06)
In patients with osteoarthritis (OA), there is a decrease in both the concentration and molecular size of hyaluronan (HA) in the synovial fluid and cartilage. Cell migration-inducing hyaluronidase 1 (CEMIP), also known as hyaluronan (HA)-binding protein involved in HA depolymerization
Jun Shiozawa et al.
The American journal of pathology, 190(5), 1046-1058 (2020-02-23)
Cell migration-inducing hyaluronidase 1 (CEMIP), also known as hyaluronan (HA)-binding protein involved in HA depolymerization (HYBID), plays a role in HA degradation. CEMIP2, also known as transmembrane protein 2 (TMEM2), possessing a sequence similarity with HYBID, is reported as a
Masayuki Shimoda et al.
The American journal of pathology, 187(5), 1162-1176 (2017-03-13)
Hyaluronan (HA) plays an important role in the development and maintenance of tissues, and its degradation is implicated in many pathologic conditions. We recently reported that HA-binding protein involved in HA depolymerization (CEMIP; alias HYBID/KIAA1199) is a key molecule in
Petra Žádníková et al.
Biomolecules, 12(2) (2022-02-26)
Hyaluronan (HA) comprises a fundamental component of the extracellular matrix and participates in a variety of biological processes. Half of the total amount of HA in the human body is present in the skin. HA exhibits a dynamic turnover; its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service