Skip to Content
Merck
All Photos(2)

Key Documents

SAB1406060

Sigma-Aldrich

Anti-KIFC3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686D23201, FLJ34694

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~77 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIFC3(3801)

General description

KIFC3 belongs to the large superfamily of kinesins, molecular motors that use the energy of ATP hydrolysis to translocate cargoes along microtubules. Members share extensive homology within a globular domain containing the microtubule- and ATP-binding sites and have a coiled-coil stalk domain that mediates oligomerization. Different kinesin family members participate in specific and diverse motile processes, such as cell division, organelle transport, and nuclear movement (Hoang et al., 1998 [PubMed 9782090]).[supplied by OMIM

Immunogen

KIFC3 (AAH34234.1, 1 a.a. ~ 687 a.a) full-length human protein.

Sequence
MVENERLRQEMRRCEAELQELRTKPAGPCPGCEHSQESAQLRDKLPQLQLEMAESKGMLSELNLEVQQKTDRLAEVELRLKDCLAEKAQEEERLSRRLRDSHETIASLRAQSPPVKYVIKTVEVESSKTKQALSESQARNQHLQEQVAMQRQVLKEMEQQLQSSHQLTARLRAQIAMYESELERAHGQMLEEMQSLEEDKNRAIEEAFARAQVEMKAVHENLAGVRTNLLTLQPALRTLTNDYNGLKRQVRGFPLLLQEALRSVKAEIGQAIEEVNSNNQELLRKYRRELQLRKKCHNELVRLKGNIRVIARVRPVTKEDGEGPEATNAVTFDADDDSIIHLLHKGKPVSFELDKVFSPQASQQDVFQEVQALVTSCIDGFNVCIFAYGQTGAGKTYTMEGTAENPGINQRALQLLFSEVQEKASDWEYTITVSAAEIYNEVLRDLLGKEPQEKLEIRLCPDGSGQLYVPGLTEFQVQSVDDINKVFEFGHTNRTTEFTNLNEHSSRSHALLIVTVRGVDCSTGLRTTGKLNLVDLAGSERVGKSGAEGSRLREAQHINKSLSALGDVIAALRSRQGHVPFRNSKLTYLLQDSLSGGSKTLMVVQVSPVEKNTSETLYSLKFAERVRSVELGPGLRRAELGSWSSQEHLEWEPACQTPQPSARAHSAPSSGTSSRPGSIRRKLQPSA

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Jun Wang et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(1), 62-69 (2013-10-12)
Previous studies identified a family of organic anion transport proteins (OATPs), many of which have C-terminal PDZ binding consensus sequences. In particular, the C-terminal four amino acids of Oatp1a1, a transporter on rat and mouse hepatocytes, comprise a consensus binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service