Skip to Content
Merck
All Photos(4)

Key Documents

SAB1405122

Sigma-Aldrich

Monoclonal Anti-SOX18 antibody produced in mouse

clone 1C4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

HLTS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~35.79 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
RAB0524RAB0574RAB0572
Gene Information

canine ... IL6(403985)

Gene Information

canine ... IL10(403628)

Gene Information

canine ... IL2(403989)

Gene Information

canine ... IL1B(403974)

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.1 ng/mL

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.1 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.4 ng/mL
standard curve range: 0.41-100 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 10.97-8000 pg/mL

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

General description

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. This protein plays a role in hair, blood vessel, and lymphatic vessel development. Mutations in this gene have been associated with recessive and dominant forms of hypotrichosis-lymphedema-telangiectasia. (provided by RefSeq)

Immunogen

SOX18 (NP_060889, 63 a.a. ~ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EPGRYGLSPAGRGERQAADESRIRRPMNAFMVWAKDERKRLAQQNPDLHNAVLSKMLGKAWKELNAAEKRPFVEEAERLRVQHLRDHP

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service