R8391
Unwire™ full racks, polyketone
Holds 24 x 30-mm tubes, blue
Synonym(s):
moulded rack, test tube rack
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
Recommended Products
material
polyketone
feature
Holds 24 x 30-mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 5970-0330
L × W × H
283 mm × 108 mm × 83 mm
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Molded of durable polyketone. Unwire test tube racks have no coating that can chip or peel and permit corrosion. Excellent replacement for wire racks; withstands most lab chemicals. Excellent thermal properties and won′t get brittle in freezers. Grid design allows tubes to be tipped from side to side for clearly marking position. These racks will not float in water baths.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service