Skip to Content
Merck
All Photos(2)

Key Documents

N3512

Sigma-Aldrich

4-Nitrophenyl α-L-arabinopyranoside

arabinosidase substrate, ≥98% (HPLC), powder

Synonym(s):

4-Nitrophenyl alpha-L-arabinopyranoside, p-Nitrophenyl alpha-L-arabinopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO7
CAS Number:
Molecular Weight:
271.22
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

4-Nitrophenyl α-L-arabinopyranoside, arabinosidase substrate

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

ethanol: water (1:1): 19.60-20.40 mg/mL

storage temp.

−20°C

SMILES string

OC1COC(Oc2ccc(cc2)N(=O)=O)C(O)C1O

InChI

1S/C11H13NO7/c13-8-5-18-11(10(15)9(8)14)19-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2

InChI key

MLJYKRYCCUGBBV-UHFFFAOYSA-N

Application

4-Nitrophenyl α-L-arabinopyranoside may be used: as a 4-nitrophenyl-glycoside substrate to study the substrate activities of TlAbf51 by determining the arabinofuranosidase (Abf) activity in standard conditions.[1] It may also be used to determine the activity of α-L-arabinofuranosidase spectrophotometrically.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Robert C Lee et al.
The Journal of biological chemistry, 278(7), 5377-5387 (2002-12-05)
An alpha-l-arabinofuranosidase and a beta-d-xylosidase, designated ARA-I and XYL, respectively, have been purified about 1,000-fold from extracts of 5-day-old barley (Hordeum vulgare L.) seedlings using ammonium sulfate fractional precipitation, ion exchange chromatography, chromatofocusing, and size-exclusion chromatography. The ARA-I has an
Tao Tu et al.
Microbial cell factories, 18(1), 138-138 (2019-08-21)
The development of sustainable technologies for plant cell wall degradation greatly depends on enzymes with hydrolytic activities against carbohydrates. The waste by-products of agricultural cereals are important biomass sources because they contain large amounts of saccharides. Achieving efficient debranching and
Robert W P Glowacki et al.
Cell host & microbe, 27(1), 79-92 (2020-01-07)
Efficient nutrient acquisition in the human gut is essential for microbial persistence. Although polysaccharides have been well-studied nutrients for the gut microbiome, other resources such as nucleic acids and nucleosides are less studied. We describe several ribose-utilization systems (RUSs) that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service