MTCC1813
MTC™ Bio SecureSeal™ Transport Tubes
size 5 mL, graduation (printed)
Synonym(s):
sample mailing tube, transportation tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear tube
conical (self standing)
polypropylene
screw cap
white cap
sterility
sterile
feature
graduation (printed)
packaging
pack of 1000 ea (10 x bags 100 ea)
manufacturer/tradename
MTC™ Bio C1813
diam. × L
16 mm × 60 mm
size
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Transport Tube, sterile, 16 x 60mm, 5ml, with screw-cap, printed graduations, self-standing, 100 tubes per bag, with caps attached, 1000/cs
Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. These self standing tubes are made of high-grade polypropylene, and include white printed graduations and marking area.
Features and Benefits
- Leak-proof, 1.25 turn screw-cap
- Meet 95kPa transportation guidelines
- Vivid, printed graduations
- Screw-caps attached Sterile, RNase, DNase, Pyrogen free
Legal Information
MTC is a trademark of MTC Bio
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service