MSST0061
SILu™Prot APOD, Apolipoprotein D human
recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled
Synonym(s):
Apo-D, ApoD
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
recombinant
expressed in HEK 293 cells
Quality Level
Assay
≥95% (SDS-PAGE)
form
lyophilized powder
potency
≥98% (Heavy amino acids incorporation efficiency by MS)
suitability
suitable for mass spectrometry (standard)
UniProt accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... APOD(347)
General description
SILu™Prot APOD is a recombinant, stable isotope-labeled human APOD which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of APOD in mass-spectrometry. SILu™Prot APOD is a protein of 189 amino acids (including a C-terminal polyhistidine and tag), with a calculated molecular mass of 21.8 kDa.
Biochem/physiol Actions
Apolipoprotein D is a member of the Apolipoprotein family. Unlike other lipoproteins, which are mainly produced in the liver, apolipoprotein D is mainly produced in the brain and testes. It was found to be a putative biomarker of androgen receptor function in androgen insensitivity syndrome, the most common cause of disorders of sex development. Apolipoprotein D is associated with neurological disorders and nerve injury, especially related to myelin sheath. It was shown to be elevated in a rat model of stroke, and is elevated in patients with schizophrenia, bipolar disorder, and Alzheimer′s disease.
Sequence
QAFHLGKCPNPPVQENFDVNKYLGRWYEIEKIPTTFENGRCIQANYSLMENGKIKVLNQELRADGTVNQIEGEATPVNLTEPAKLEVKFSWFMPSAPYWILATDYENYALVYSCTCIIQLFHVDFAWILARNPNLPPETVDSLKNILTSNNIDVKKMTVTDQVNCPKLSDYKDDDDKGHHHHHHHHGGQ
Physical form
Supplied as a lyophilized powder containing phosphate buffered saline.
Legal Information
This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.
SILu is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service