MAK495
Aspartate Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Aspartate Quantification Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
Quality Level
application(s)
pharmaceutical
detection method
colorimetric
fluorometric
relevant disease(s)
endocrinological disorders, diabetes; neurological disorders
shipped in
wet ice
storage temp.
2-8°C
Related Categories
General description
Aspartate, a nonessential amino acid, is a precursor to several other amino acids and is an excitatory neurotransmitter. Aspartate is involved in the urea cycle and gluconeogenesis.
Features and Benefits
- Linear detection range: 2 to 400 µM aspartate for the colorimetric assay and 1 to 50 µM for the fluorometric assay.
Suitability
The kit is suitable for aspartate determination in plasma, serum, tissue and culture media, as well as for studying the effects of drugs on aspartate metabolism.
Principle
In the Aspartate Assay Kit, aspartate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity of the oxidized dye at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the aspartate concentration in the sample.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service