HS120203
Microtube Storage Box
yellow polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
All Photos(2)
About This Item
UNSPSC Code:
41123300
NACRES:
NB.22
Recommended Products
material
yellow polypropylene
feature
Holds 100 x 1.5-2.0 mL tubes
packaging
pk of 5
manufacturer/tradename
Heathrow Scientific HS120203
well diam.
12.3 mm
Related Categories
Features and Benefits
- for larger storage requirements of 1.5-2.0 mL microtubes or cryogenic vials
- easy-open hinged lid with snap clasp
- autoclavable
- designed for quick visual identification
- uniform wells with location indentification
- tube capacity is indicated on the outside of the box
- translucent vibrant colors with frosted labeling area
- debossed with a distinctive wave design
Choose from one of the most recent versions:
Certificates of Analysis (COA)