Skip to Content
Merck
All Photos(1)

Key Documents

HS120000

eCount colony counter with pen, white and blue

Synonym(s):

electronic colony counter, manual colony counter aid, microbial colony counter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

Pricing and availability is not currently available.

manufacturer/tradename

Heathrow Scientific HS120000

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PR0G000T3PR0G000S2PR0G000S1
manufacturer/tradename

Progard®

manufacturer/tradename

Progard®

manufacturer/tradename

Progard®

manufacturer/tradename

Progard®

feature

Optimal protection of Reverse Osmosis (RO) membrane, optimized system lifetime and performance. Removal of large particles, organic compounds and free chlorine from feed water with a silt density index (SDI) up to 12

feature

Optimal protection of RO membrane, optimized system lifetime and performance, feed water with a silt density index up to 5.

feature

Optimal protection of Reverse Osmosis (RO) membrane, optimized system lifetime and performance. Removal of large particles, organic compounds and free chlorine from feed water with a silt density index (SDI) up to 12.

feature

-

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 1 unit

packaging

pkg of 1 unit

material

EPDM rubber

material

-

material

EPDM rubber , polypropylene

material

polypropylene (and EPDM)

General description

The Rapid, accurate eCount colony counter combines a multi-function electronic counter with a Sharpie pen for marking to prevent missing or double-counting colonies.

  • Light, ergonomic body is balanced for easy manipulation
  • Select between an audible beep or an led flash to acknowledge count
  • Store up to 32 separate counts and display the total of all counts
  • Count up/down verification
  • Quick and easy to change between different colored markers to check different colony counts in the same view
  • Lock counter off to allow use of the marker without registering more counts
  • US Patent No. D564385 and EP Registered Design No. 705751-0001

Other Notes

Package Includes:
  • eCount colony counter
  • Black Sharpie pen for highly visible marks on both plastic and glass
  • Long-life button battery
  • Hand tally adapter counts without marking
  • Weighted stand for convenient bench top use


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service