Skip to Content
Merck
All Photos(6)

Key Documents

HPA038421

Sigma-Aldrich

Anti-WDR48 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA1449, Anti-P80, Anti-SPG60

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

SIIQCHILNDKRHILTKDTNNNVAYWDVLKACKVEDLGKVDFEDEIKKRFKMVYVPNWFSVDLKTGMLTITLDESDCFAAWVSAKDAGF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDR48(57599)

General description

The gene WDR48 (WD repeat domain 48) is mapped to human chromosome 3p22.2. It is a cofactor for deubiquitinating enzymes USP1 (ubiquitin specific peptidase 1), USP12 and USP46.

Immunogen

WD repeat domain 48 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-WDR48 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

USP1-WDR48 (ubiquitin specific peptidase 1 - WD repeat domain 48) complex is involved in DNA damage responses. It is responsible for repair of interstrand DNA crosslinks through the Fanconi anemia pathway. It also reverses monoubiquitination of PCNA (proliferating cell nuclear antigen) and regulates translesion synthesis, a DNA damage tolerance response. This complex also participates in homologous recombination. USP1-WDR48 complex mainly controls nuclear protein ubiquitination. WDR48 also works as a tumor suppressor by enhancing PHLPP1 (PH domain leucine-rich repeat protein phosphatase 1) stability. The WDR48 - USP12 complex is responsible for deubiquitination of PHLPP1, which is needed for tumor suppressor function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79337

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The EBNA3 family of Epstein-Barr virus nuclear proteins associates with the USP46/USP12 deubiquitination complexes to regulate lymphoblastoid cell line growth.
Ohashi M, et al.
PLoS Pathogens, 11, e1004822-e1004822 (2015)
Delving into the complexity of hereditary spastic paraplegias: how unexpected phenotypes and inheritance modes are revolutionizing their nosology.
Tesson C, et al.
Human Genetics, 134, 511-538 (2015)
WD repeat protein WDR48 in complex with deubiquitinase USP12 suppresses Akt-dependent cell survival signaling by stabilizing PH domain leucine-rich repeat protein phosphatase 1 (PHLPP1).
Gangula NR and Maddika S
The Journal of Biological Chemistry, 288, 34545-34554 (2013)
Mutations in the 'Fingers' subdomain of the deubiquitinase USP1 modulate its function and activity.
Olazabal-Herrero A, et al.
FEBS Journal, 283, 929-946 (2016)
Makoto Ohashi et al.
PLoS pathogens, 11(4), e1004822-e1004822 (2015-04-10)
The Epstein-Barr virus (EBV) nuclear proteins EBNA3A, EBNA3B, and EBNA3C interact with the cell DNA binding protein RBPJ and regulate cell and viral genes. Repression of the CDKN2A tumor suppressor gene products p16(INK4A) and p14(ARF) by EBNA3A and EBNA3C is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service